Assay Details
Target Gene Details
Entrez Gene ID: | 55388 |
Gene Name: | minichromosome maintenance 10 replication initiation factor |
Gene Aliases: |
CNA43, DNA43, PRO2249 |
Location: |
Chr.10:13161554-13211110 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM10 | NM_018518.4 | NP_060988.3 | ||
| NM_182751.2 | NP_877428.1 | |||
| XM_011519538.2 | XP_011517840.1 | |||
| AB042719.1 | BAB18723.1 | |||
| AK055695.1 | BAB70988.1 | |||
| AK292701.1 | ||||
| AL136840.1 | CAB66774.2 | |||
| BC101727.1 | AAI01728.1 | |||
| BC143490.1 | ||||
| DA024424.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76796379] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map