Assay Details
Target Gene Details
Entrez Gene ID: | 80314 |
Gene Name: | enhancer of polycomb homolog 1 |
Gene Aliases: |
Epl1 |
Location: |
Chr.10:32267716-32378798 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPC1 | NM_001282391.1 | NP_001269320.1 | ||
| XM_006717510.1 | XP_006717573.1 | |||
| AF286905.1 | AAK60501.1 | |||
| AK299676.1 | ||||
| AK309309.1 | ||||
| DB070690.1 |
Target Gene Details
Entrez Gene ID: | 102031319 |
Gene Name: | uncharacterized LOC102031319 |
Gene Aliases: |
- |
Location: |
Chr.10:32347364-32374488 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102031319 | NR_104163.1 | |||
| BX107541.1 | ||||
| BX118505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3622828 | Chr.10:32351820 - 32356489 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map