Assay Details
Target Gene Details
Entrez Gene ID: | 6865 |
Gene Name: | tachykinin receptor 2 |
Gene Aliases: |
NK2R, NKNAR, SKR, TAC2R |
Location: |
Chr.10:69404202-69416918 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TACR2 | NM_001057.2 | NP_001048.2 | ||
| AK096906.1 | ||||
| AK291996.1 | ||||
| AK300649.1 | ||||
| AY322545.1 | AAP84358.1 | |||
| BC096842.1 | AAH96842.1 | |||
| FJ200479.1 | ||||
| M57414.1 | AAC31760.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551453 | Chr.10:69384195 - 69415853 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map