Assay Details
Target Gene Details
Entrez Gene ID: | 8861 |
Gene Name: | LIM domain binding 1 |
Gene Aliases: |
CLIM-2, CLIM2, LDB-1, NLI |
Location: |
Chr.10:102102100-102120473 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDB1 | NM_001113407.2 | NP_001106878.1 | ||
| NM_001321612.1 | NP_001308541.1 | |||
| XM_017016867.1 | XP_016872356.1 | |||
| XM_017016868.1 | XP_016872357.1 | |||
| CB988590.1 | ||||
| CV575131.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76495526] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map