Assay Details
Target Gene Details
Entrez Gene ID: | 9188 |
Gene Name: | DEAD-box helicase 21 |
Gene Aliases: |
GUA, GURDB, RH-II/GU, RH-II/GuA |
Location: |
Chr.10:68956123-68985069 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX21 | NM_001256910.1 | NP_001243839.1 | ||
| NM_004728.3 | NP_004719.2 | |||
| XM_011540336.1 | XP_011538638.1 | |||
| XM_017016910.1 | XP_016872399.1 | |||
| AK315585.1 | ||||
| BC008071.2 | AAH08071.2 | |||
| BX648405.1 | ||||
| CR749598.1 | CAH18395.1 | |||
| U41387.1 | AAB02546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519999 | Chr.10:68931952 - 69015325 on Build GRCh38 | Loss |
|
| nsv831907 | Chr.10:68871974 - 69029649 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map