Assay Details
Target Gene Details
Entrez Gene ID: | 1793 |
Gene Name: | dedicator of cytokinesis 1 |
Gene Aliases: |
DOCK180, ced5 |
Location: |
Chr.10:126905409-127452517 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK1 | NM_001290223.1 | NP_001277152.1 | ||
| NM_001380.4 | NP_001371.1 | |||
| XM_017015815.1 | XP_016871304.1 | |||
| BC146857.1 | ||||
| BX482033.1 | ||||
| D50857.1 | BAA09454.1 | |||
| DA350224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045509 | Chr.10:126162634 - 127193365 on Build GRCh38 | Gain |
|
| esv3891896 | Chr.10:126281817 - 127117455 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78853408] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map