Assay Details
Target Gene Details
Entrez Gene ID: | 83643 |
Gene Name: | coiled-coil domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.10:12896625-13099773 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC3 | NM_001282658.1 | NP_001269587.1 | ||
| NM_031455.3 | NP_113643.1 | |||
| AK095792.1 | ||||
| AK307203.1 | ||||
| AK307304.1 | ||||
| AL136562.1 | CAB66497.1 | |||
| BC051334.1 | AAH51334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053098 | Chr.10:12904089 - 12926547 on Build GRCh38 | Gain |
|

Genomic Map