Assay Details
Target Gene Details
Entrez Gene ID: | 27291 |
Gene Name: | R3H domain and coiled-coil containing 1 like |
Gene Aliases: |
C10orf28, GIDRP86, GIDRP88, PSORT |
Location: |
Chr.10:98134608-98244897 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3303473 | Chr.10:98137968 - 98137969 on Build GRCh38 | Mobile Element Insertion |
|
| esv3392688 | Chr.10:98137958 - 98138000 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map