Assay Details
Target Gene Details
Entrez Gene ID: | 1559 |
Gene Name: | cytochrome P450 family 2 subfamily C member 9 |
Gene Aliases: |
CPC9, CYP2C, CYP2C10, CYPIIC9, P450IIC9 |
Location: |
Chr.10:94905867-94989395 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2C9 | NM_000771.3 | NP_000762.2 | ||
| XM_017015758.1 | XP_016871247.1 | |||
| AK289420.1 | ||||
| AK298458.1 | ||||
| BC125054.1 | ||||
| D00173.1 | BAA00123.1 | |||
| M15331.1 | AAA52157.1 | |||
| M21939.1 | AAA52158.1 | |||
| M21940.1 | AAA52159.1 | |||
| M61855.1 | ||||
| M61857.1 | ||||
| S46963.1 | AAB23864.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624264 | Chr.10:94923968 - 95099386 on Build GRCh38 | Gain |
|
| esv2741937 | Chr.10:94845829 - 94984654 on Build GRCh38 | Deletion |
|
| nsv551962 | Chr.10:94920508 - 94974582 on Build GRCh38 | Loss |
|
| nsv1035409 | Chr.10:94735420 - 95072300 on Build GRCh38 | Loss |
|
| esv3624265 | Chr.10:94931890 - 95004479 on Build GRCh38 | Gain |
|
| nsv551963 | Chr.10:94960505 - 94984186 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map