Assay Details
Target Gene Details
Entrez Gene ID: | 64081 |
Gene Name: | phenazine biosynthesis like protein domain containing |
Gene Aliases: |
HEL-S-306, MAWBP, MAWDBP |
Location: |
Chr.10:68282660-68332958 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PBLD | NM_001033083.1 | NP_001028255.1 | ||
| NM_022129.3 | NP_071412.2 | |||
| XM_005270028.3 | XP_005270085.1 | |||
| XM_011540060.2 | XP_011538362.1 | |||
| XM_017016513.1 | XP_016872002.1 | |||
| XM_017016514.1 | XP_016872003.1 | |||
| AK027673.1 | BAB55285.1 | |||
| BC009738.1 | AAH09738.1 | |||
| CB121561.1 | ||||
| GQ472233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv437125 | Chr.10:68303779 - 68315812 on Build GRCh38 | Loss |
|
| esv3623712 | Chr.10:68308472 - 68310892 on Build GRCh38 | Loss |
|
| esv2738017 | Chr.10:68308461 - 68310890 on Build GRCh38 | Deletion |
|
| nsv517615 | Chr.10:68304752 - 68309476 on Build GRCh38 | Loss |
|
| esv3546453 | Chr.10:68308443 - 68310930 on Build GRCh38 | Deletion |
|
| dgv158n67 | Chr.10:68308696 - 68310424 on Build GRCh38 | Loss |
|
| dgv150e199 | Chr.10:68308468 - 68310887 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map