Assay Details
Target Gene Details
Entrez Gene ID: | 5588 |
Gene Name: | protein kinase C theta |
Gene Aliases: |
PRKCT, nPKC-theta |
Location: |
Chr.10:6393038-6580646 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCQ | NM_001323267.1 | NP_001310196.1 | ||
| XM_005252497.4 | XP_005252554.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73621305] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map