Assay Details
Target Gene Details
Entrez Gene ID: | 27291 |
Gene Name: | R3H domain and coiled-coil containing 1 like |
Gene Aliases: |
C10orf28, GIDRP86, GIDRP88, PSORT |
Location: |
Chr.10:98134608-98244897 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
More Information
Set Membership: |
|

Genomic Map