Assay Details
Target Gene Details
Entrez Gene ID: | 64215 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C1 |
Gene Aliases: |
DNAJL1, ERdj1, HTJ1, MTJ1 |
Location: |
Chr.10:21756548-22003768 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC1 | NM_022365.3 | NP_071760.2 | ||
| XM_011519614.2 | XP_011517916.1 | |||
| AF427142.1 | AAL24046.1 | |||
| AK026062.1 | BAB15343.1 | |||
| AK027263.1 | BAB55004.1 | |||
| AY225122.1 | AAP50497.1 | |||
| BC110894.1 | AAI10895.1 | |||
| BC139752.1 |
More Information
Set Membership: |
|

Genomic Map