Assay Details
Target Gene Details
Entrez Gene ID: | 10579 |
Gene Name: | transforming acidic coiled-coil containing protein 2 |
Gene Aliases: |
AZU-1, ECTACC |
Location: |
Chr.10:121989174-122254545 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TACC2 | NM_001291876.1 | NP_001278805.1 | ||
| NM_001291877.1 | NP_001278806.1 | |||
| NM_206861.2 | NP_996743.1 | |||
| NM_206862.3 | NP_996744.3 | |||
| AF528098.1 | AAO62629.1 | |||
| AF528099.1 | AAO62630.1 | |||
| BC144599.1 | ||||
| BC144600.1 | ||||
| BC150627.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045815 | Chr.10:121986932 - 122086108 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map