Assay Details
Target Gene Details
Entrez Gene ID: | 2665 |
Gene Name: | GDP dissociation inhibitor 2 |
Gene Aliases: |
HEL-S-46e, RABGDIB |
Location: |
Chr.10:5765222-5813549 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDI2 | NM_001115156.1 | NP_001108628.1 | ||
| NM_001494.3 | NP_001485.2 | |||
| XM_017016071.1 | XP_016871560.1 | |||
| AB451295.1 | ||||
| AB451429.1 | ||||
| AF144713.1 | AAD34588.1 | |||
| AK297177.1 | ||||
| AK297554.1 | ||||
| AK309360.1 | ||||
| AK312485.1 | ||||
| BC005145.2 | AAH05145.1 | |||
| BT006868.1 | AAP35514.1 | |||
| BX648038.1 | ||||
| CR457073.1 | CAG33354.1 | |||
| D13988.1 | BAA03095.1 | |||
| EU794614.1 | ||||
| Y13286.1 | CAA73734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515935 | Chr.10:5778431 - 5827876 on Build GRCh38 | Gain |
|
| nsv831775 | Chr.10:5762372 - 5958886 on Build GRCh38 | Loss |
|
| dgv1e49 | Chr.10:5684858 - 6183224 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map