Assay Details
Target Gene Details
Entrez Gene ID: | 84287 |
Gene Name: | zinc finger DHHC-type containing 16 |
Gene Aliases: |
APH2, DHHC-16 |
Location: |
Chr.10:97446131-97457370 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
More Information
Set Membership: |
|

Genomic Map