Assay Details
Target Gene Details
Entrez Gene ID: | 9562 |
Gene Name: | multiple inositol-polyphosphate phosphatase 1 |
Gene Aliases: |
HIPER1, MINPP2, MIPP |
Location: |
Chr.10:87502792-87553461 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MINPP1 | NM_001178117.1 | NP_001171588.1 | ||
| NM_001178118.1 | NP_001171589.1 | |||
| NM_004897.4 | NP_004888.2 | |||
| XM_006718078.2 | XP_006718141.1 | |||
| XM_011540379.2 | XP_011538681.1 | |||
| XM_017016966.1 | XP_016872455.1 | |||
| AB209819.1 | BAD93056.1 | |||
| AF046914.1 | AAD02437.1 | |||
| AF084943.1 | AAD09751.1 | |||
| AF084944.1 | AAD09752.1 | |||
| AK304624.1 | ||||
| AK309176.1 | ||||
| AK312937.1 | ||||
| AL050356.1 | CAB43673.1 | |||
| AY358938.1 | AAQ89297.1 | |||
| BC032504.1 | AAH32504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891880 | Chr.10:87343291 - 87725505 on Build GRCh38 | Gain |
|
| nsv511458 | Chr.10:87513822 - 87532880 on Build GRCh38 | Loss |
|
| nsv428236 | Chr.10:86866859 - 87550005 on Build GRCh38 | Loss |
|
| nsv508598 | Chr.10:87521049 - 87618215 on Build GRCh38 | Deletion |
|
| esv2759774 | Chr.10:86755301 - 87550005 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map