Assay Details
Target Gene Details
Entrez Gene ID: | 8861 |
Gene Name: | LIM domain binding 1 |
Gene Aliases: |
CLIM-2, CLIM2, LDB-1, NLI |
Location: |
Chr.10:102102100-102120473 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDB1 | NM_001113407.2 | 11 | 3049 | NP_001106878.1 |
| NM_001321612.1 | 11 | 3043 | NP_001308541.1 | |
| XM_017016867.1 | XP_016872356.1 | |||
| XM_017016868.1 | XP_016872357.1 | |||
| AW137421.1 |
More Information
Set Membership: |
|

Genomic Map