Assay Details
Target Gene Details
Entrez Gene ID: | 100507633 |
Gene Name: | TNKS2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.10:91782839-91798291 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNKS2-AS1 | NR_108070.1 | 2 | 436 | |
| NR_108071.1 | 2 | 107 | ||
| AA776692.1 | 1 | 273 | ||
| BM678746.1 | 2 | 436 |

Genomic Map