Assay Details
Target Gene Details
Entrez Gene ID: | 11101 |
Gene Name: | arginyltransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.10:121740421-121928801 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATE1 | NM_001001976.2 | NP_001001976.1 | ||
| NM_001288734.1 | NP_001275663.1 | |||
| NM_001288735.1 | NP_001275664.1 | |||
| NM_001288736.1 | NP_001275665.1 | |||
| NM_007041.3 | NP_008972.2 | |||
| NR_110006.1 | ||||
| XM_005269458.2 | XP_005269515.1 | |||
| XM_005269459.3 | XP_005269516.1 | |||
| XM_017015597.1 | XP_016871086.1 | |||
| XM_017015598.1 | XP_016871087.1 | |||
| XM_017015599.1 | XP_016871088.1 | |||
| XM_017015600.1 | XP_016871089.1 | |||
| XM_017015602.1 | XP_016871091.1 | |||
| XM_017015604.1 | XP_016871093.1 | |||
| AF079098.1 | AAD12366.1 | |||
| AF079099.1 | AAD12367.1 | |||
| AK124669.1 | ||||
| AK292657.1 | ||||
| AK295642.1 | ||||
| AK296356.1 | ||||
| AK299101.1 | ||||
| AK303604.1 | ||||
| AK308926.1 | ||||
| AK313057.1 | ||||
| BC022026.1 | AAH22026.2 |
More Information
Additional Information:
For this assay, SNP(s) [rs77376812] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map