Assay Details
Target Gene Details
Entrez Gene ID: | 355 |
Gene Name: | Fas cell surface death receptor |
Gene Aliases: |
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6 |
Location: |
Chr.10:88968429-89017061 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831943 | Chr.10:88905079 - 89106077 on Build GRCh38 | Gain |
|
| esv3624144 | Chr.10:88876937 - 89011636 on Build GRCh38 | Gain |
|
| esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map