Assay Details
Target Gene Details
Entrez Gene ID: | 3189 |
Gene Name: | heterogeneous nuclear ribonucleoprotein H3 |
Gene Aliases: |
2H9, HNRPH3 |
Location: |
Chr.10:68331174-68343196 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPH3 | XM_017016168.1 | 1 | 425 | XP_016871657.1 |
| XM_017016169.1 | 1 | 425 | XP_016871658.1 | |
| AK091411.1 | 1 | 425 |
Target Gene Details
Entrez Gene ID: | 64081 |
Gene Name: | phenazine biosynthesis like protein domain containing |
Gene Aliases: |
HEL-S-306, MAWBP, MAWDBP |
Location: |
Chr.10:68282660-68332958 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PBLD | NM_001033083.1 | NP_001028255.1 | ||
| NM_022129.3 | NP_071412.2 | |||
| XM_005270028.3 | XP_005270085.1 | |||
| XM_011540060.2 | XP_011538362.1 | |||
| XM_017016513.1 | XP_016872002.1 | |||
| XM_017016514.1 | XP_016872003.1 | |||
| AK027673.1 | BAB55285.1 | |||
| BC009738.1 | AAH09738.1 | |||
| CB121561.1 | ||||
| GQ472233.1 |
More Information
Set Membership: |
|

Genomic Map