Assay Details
Target Gene Details
Entrez Gene ID: | 7511 |
Gene Name: | X-prolyl aminopeptidase 1 |
Gene Aliases: |
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1 |
Location: |
Chr.10:109864766-109923553 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XPNPEP1 | NM_001167604.1 | NP_001161076.1 | ||
| NM_001324128.1 | NP_001311057.1 | |||
| NM_001324131.1 | NP_001311060.1 | |||
| NM_001324132.1 | NP_001311061.1 | |||
| NM_001324133.1 | NP_001311062.1 | |||
| NM_001324134.1 | NP_001311063.1 | |||
| NM_001324135.1 | NP_001311064.1 | |||
| NM_001324136.1 | NP_001311065.1 | |||
| NM_020383.3 | NP_065116.3 | |||
| XM_011540134.1 | XP_011538436.1 | |||
| XM_017016613.1 | XP_016872102.1 | |||
| XM_017016614.1 | XP_016872103.1 | |||
| XM_017016615.1 | XP_016872104.1 | |||
| AF195530.1 | AAF97866.1 | |||
| AF272981.1 | AAF75795.1 | |||
| AK093693.1 | ||||
| AK095447.1 | 1 | 825 | ||
| AK223513.1 | BAD97233.1 | |||
| AK289436.1 | ||||
| AL833411.1 | CAD38640.1 | |||
| BC005126.2 | AAH05126.1 | |||
| BC007579.1 | AAH07579.1 | |||
| BC013417.2 | AAH13417.4 | |||
| BX537682.1 | 11 | 2450 | CAH18706.2 | |
| CR456922.1 | CAG33203.1 | |||
| X95762.1 | CAA65068.1 |
More Information
Set Membership: |
|

Genomic Map