Assay Details
Target Gene Details
Entrez Gene ID: | 80314 |
Gene Name: | enhancer of polycomb homolog 1 |
Gene Aliases: |
Epl1 |
Location: |
Chr.10:32267716-32378798 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPC1 | NM_001272004.1 | NP_001258933.1 | ||
| NM_001272019.2 | NP_001258948.1 | |||
| NM_001282391.1 | NP_001269320.1 | |||
| NM_025209.3 | NP_079485.1 | |||
| XM_006717510.1 | XP_006717573.1 | |||
| AF277374.1 | AAG41402.1 | |||
| AF286905.1 | AAK60501.1 | |||
| AK024329.1 | BAB14888.1 | |||
| AK092304.1 | BAC03857.1 | |||
| AK299676.1 | ||||
| AK309309.1 | ||||
| AK309822.1 | ||||
| BC036529.1 | AAH36529.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114138328] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map