Assay Details
Target Gene Details
Entrez Gene ID: | 55315 |
Gene Name: | solute carrier family 29 member 3 |
Gene Aliases: |
ENT3, HCLAP, HJCD, PHID |
Location: |
Chr.10:71319253-71363390 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC29A3 | NM_001174098.1 | NP_001167569.1 | ||
| NM_018344.5 | NP_060814.4 | |||
| NR_033413.1 | ||||
| NR_033414.1 | ||||
| XM_006717910.3 | XP_006717973.1 | |||
| XM_017016377.1 | XP_016871866.1 | |||
| XM_017016378.1 | XP_016871867.1 | |||
| AF326987.1 | AAK00958.1 | |||
| AK002022.1 | BAA92041.1 | |||
| AK304503.1 | ||||
| AK314497.1 | ||||
| AK316152.1 | ||||
| AX775915.1 | CAE11657.1 | |||
| AX775917.1 | CAE11658.1 | |||
| AX775919.1 | CAE11659.1 | |||
| AX775921.1 | CAE11660.1 | |||
| AY288928.1 | AAP41133.1 | |||
| AY358686.1 | AAQ89049.1 | |||
| BC041575.1 | AAH41575.1 | |||
| BC120996.1 | ||||
| BC120997.1 | ||||
| DC319920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044971 | Chr.10:71272698 - 71673682 on Build GRCh38 | Gain |
|
| nsv467371 | Chr.10:71302291 - 71340509 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map