Assay Details
Target Gene Details
Entrez Gene ID: | 5950 |
Gene Name: | retinol binding protein 4 |
Gene Aliases: |
MCOPCB10, RDCCAS |
Location: |
Chr.10:93591836-93601744 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBP4 | NM_001323517.1 | NP_001310446.1 | ||
NM_001323518.1 | NP_001310447.1 | |||
NM_006744.3 | NP_006735.2 | |||
BC020633.1 | AAH20633.1 | |||
BG565176.1 | ||||
BI712834.1 | ||||
X00129.1 | CAA24959.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
