Assay Details
Target Gene Details
Entrez Gene ID: | 79004 |
Gene Name: | CUE domain containing 2 |
Gene Aliases: |
C10orf66, bA18I14.5 |
Location: |
Chr.10:102423245-102432666 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CUEDC2 | NM_024040.2 | NP_076945.2 | ||
| BC000262.1 | AAH00262.1 | |||
| BM756951.1 | ||||
| CX165866.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75659956] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map