Assay Details
Target Gene Details
Entrez Gene ID: | 22852 |
Gene Name: | ankyrin repeat domain 26 |
Gene Aliases: |
THC2, bA145E8.1 |
Location: |
Chr.10:26938195-27100498 on Build GRCh38 |
Assay Gene Location: | Within Exon 46 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD26 | XM_017015929.1 | 41 | 21967 | XP_016871418.1 |
Target Gene Details
Entrez Gene ID: | 387644 |
Gene Name: | long intergenic non-protein coding RNA 202-1 |
Gene Aliases: |
C10orf51, LINC00202, NCRNA00202, bB27G4.1 |
Location: |
Chr.10:26931206-26942001 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00202-1 | NR_026795.1 | 1 | 779 | |
| AK128027.1 | 1 | 1469 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831812 | Chr.10:26772577 - 26945338 on Build GRCh38 | Gain |
|
| dgv695n100 | Chr.10:26936975 - 26957892 on Build GRCh38 | Gain |
|
| esv3415799 | Chr.10:26640952 - 26944612 on Build GRCh38 | Duplication |
|
| esv2750888 | Chr.10:26936718 - 26952851 on Build GRCh38 | Gain |
|
| nsv947784 | Chr.10:26935691 - 26946921 on Build GRCh38 | Duplication |
|
| esv2664616 | Chr.10:26927640 - 27310597 on Build GRCh38 | Deletion |
|
| nsv1161935 | Chr.10:26934974 - 26951503 on Build GRCh38 | Duplication |
|

Genomic Map