Assay Details
Target Gene Details
Entrez Gene ID: | 59338 |
Gene Name: | pleckstrin homology domain containing A1 |
Gene Aliases: |
TAPP1 |
Location: |
Chr.10:122374578-122432355 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHA1 | NM_001001974.2 | NP_001001974.1 | ||
| XM_005270016.1 | XP_005270073.1 | |||
| XM_005270019.1 | XP_005270076.1 | |||
| XM_005270020.1 | XP_005270077.1 | |||
| XM_005270021.4 | XP_005270078.1 | |||
| XM_011540017.1 | 1 | 115 | XP_011538319.1 | |
| XM_011540018.1 | 1 | 167 | XP_011538320.1 | |
| XM_011540021.1 | XP_011538323.1 | |||
| XM_011540022.1 | 1 | 143 | XP_011538324.1 | |
| XM_017016478.1 | XP_016871967.1 | |||
| XM_017016479.1 | XP_016871968.1 | |||
| XM_017016480.1 | 1 | 167 | XP_016871969.1 | |
| XM_017016481.1 | 1 | 115 | XP_016871970.1 | |
| XM_017016482.1 | 1 | 143 | XP_016871971.1 | |
| XM_017016483.1 | 1 | 167 | XP_016871972.1 | |
| XM_017016484.1 | 1 | 115 | XP_016871973.1 | |
| XM_017016485.1 | XP_016871974.1 | |||
| XM_017016486.1 | 1 | 115 | XP_016871975.1 | |
| XM_017016487.1 | 1 | 167 | XP_016871976.1 | |
| XM_017016488.1 | XP_016871977.1 | |||
| XM_017016489.1 | XP_016871978.1 | |||
| XM_017016490.1 | 1 | 167 | XP_016871979.1 | |
| XM_017016491.1 | XP_016871980.1 | |||
| AK314660.1 | ||||
| BC001136.2 | AAH01136.1 | |||
| CN310748.1 | ||||
| DA923219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042640 | Chr.10:122369174 - 122442149 on Build GRCh38 | Loss |
|
| nsv1069520 | Chr.10:122374483 - 122375384 on Build GRCh38 | Deletion |
|
| nsv7578 | Chr.10:122339098 - 122384546 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map