Assay Details
Target Gene Details
Entrez Gene ID: | 219749 |
Gene Name: | zinc finger protein 25 |
Gene Aliases: |
KOX19, Zfp9 |
Location: |
Chr.10:37949573-37976656 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF25 | XM_005252384.2 | 7 | 4140 | XP_005252441.1 |
| XM_005252385.2 | 7 | 4100 | XP_005252442.1 | |
| XM_005252386.2 | 6 | 4447 | XP_005252443.1 | |
| XM_011519352.2 | 7 | 4074 | XP_011517654.1 | |
| XM_011519354.2 | 8 | 4455 | XP_011517656.1 | |
| XM_011519355.2 | 7 | 4511 | XP_011517657.1 | |
| XM_011519356.2 | 8 | 4253 | XP_011517658.1 | |
| XM_017015837.1 | 7 | 4470 | XP_016871326.1 | |
| XM_017015838.1 | 7 | 4684 | XP_016871327.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040704 | Chr.10:37651096 - 38162989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map