Assay Details
Target Gene Details
Entrez Gene ID: | 142891 |
Gene Name: | sterile alpha motif domain containing 8 |
Gene Aliases: |
HEL-177, SMSr |
Location: |
Chr.10:75099592-75182123 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMD8 | NM_001174156.1 | NP_001167627.1 | ||
| NM_144660.2 | NP_653261.1 | |||
| XM_005269541.4 | XP_005269598.1 | |||
| XM_011539311.1 | XP_011537613.1 | |||
| XM_011539312.2 | XP_011537614.1 | |||
| AK057811.1 | BAB71586.1 | |||
| BC080593.1 | AAH80593.1 | |||
| EU668332.1 | ||||
| GQ472208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831913 | Chr.10:75101265 - 75261737 on Build GRCh38 | Loss |
|
| dgv927n100 | Chr.10:75024700 - 75329039 on Build GRCh38 | Gain |
|
| nsv7199 | Chr.10:75077582 - 75112320 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map