Assay Details
Target Gene Details
Entrez Gene ID: | 1559 |
Gene Name: | cytochrome P450 family 2 subfamily C member 9 |
Gene Aliases: |
CPC9, CYP2C, CYP2C10, CYPIIC9, P450IIC9 |
Location: |
Chr.10:94905867-94989395 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2C9 | XM_017015758.1 | XP_016871247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624263 | Chr.10:94900820 - 94910519 on Build GRCh38 | Gain |
|
| nsv523259 | Chr.10:94774827 - 94933970 on Build GRCh38 | Gain |
|
| esv2741937 | Chr.10:94845829 - 94984654 on Build GRCh38 | Deletion |
|
| nsv1047782 | Chr.10:94821337 - 94958722 on Build GRCh38 | Gain |
|
| esv2674280 | Chr.10:94810345 - 94966153 on Build GRCh38 | Deletion |
|
| esv3891886 | Chr.10:94694399 - 94965778 on Build GRCh38 | Loss |
|
| dgv1355n54 | Chr.10:94684492 - 94930614 on Build GRCh38 | Loss |
|
| nsv1035409 | Chr.10:94735420 - 95072300 on Build GRCh38 | Loss |
|
| esv2739707 | Chr.10:94680312 - 94940071 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map