Assay Details
Target Gene Details
Entrez Gene ID: | 7581 |
Gene Name: | zinc finger protein 33A |
Gene Aliases: |
KOX2, KOX31, KOX5, NF11A, ZNF11, ZNF11A, ZNF33, ZZAPK |
Location: |
Chr.10:38010483-38067834 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF33A | NR_136709.1 | |||
| NR_136710.1 | ||||
| AK027057.1 | BAB15642.1 | |||
| BC030511.1 | AAH30511.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040704 | Chr.10:37651096 - 38162989 on Build GRCh38 | Gain |
|
| esv3576438 | Chr.10:38002320 - 38607542 on Build GRCh38 | Gain |
|
| nsv550419 | Chr.10:38022849 - 38212726 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map