Assay Details
Target Gene Details
Entrez Gene ID: | 84457 |
Gene Name: | phytanoyl-CoA 2-hydroxylase interacting protein like |
Gene Aliases: |
- |
Location: |
Chr.10:59174811-59247774 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHYHIPL | XM_011540276.2 | 1 | 959 | XP_011538578.1 |
| XM_017016783.1 | 1 | 959 | XP_016872272.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34715 | Chr.10:59096134 - 59179302 on Build GRCh38 | Gain |
|
| esv2759761 | Chr.10:58958273 - 59211601 on Build GRCh38 | Gain |
|

Genomic Map