Assay Details
Target Gene Details
Entrez Gene ID: | 56521 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C12 |
Gene Aliases: |
JDP1 |
Location: |
Chr.10:67796665-67838179 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC12 | NM_021800.2 | NP_068572.1 | ||
| XM_017016431.1 | XP_016871920.1 | |||
| XM_017016432.1 | XP_016871921.1 | |||
| AF176012.1 | AAD52650.1 | |||
| AK289537.1 | ||||
| BC017018.1 | AAH17018.1 | |||
| CR457184.1 | CAG33465.1 |
More Information
Set Membership: |
|

Genomic Map