Assay Details
Target Gene Details
Entrez Gene ID: | 85450 |
Gene Name: | inositol 1,4,5-trisphosphate receptor interacting protein |
Gene Aliases: |
DANGER, KIAA1754, bA127L20, bA127L20.2 |
Location: |
Chr.10:104309696-104338493 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPRIP | NM_001272012.1 | NP_001258941.1 | ||
| NM_001272013.1 | NP_001258942.1 | |||
| NM_033397.3 | NP_203755.1 | |||
| AB051541.1 | ||||
| AK056526.1 | BAB71204.1 | |||
| AK129877.1 | ||||
| BC040536.1 | AAH40536.1 | |||
| BC070108.1 | AAH70108.1 | |||
| BX436559.2 | ||||
| DA416329.1 | ||||
| DA448042.1 | ||||
| DB215055.1 |
Target Gene Details
Entrez Gene ID: | 101927472 |
Gene Name: | uncharacterized LOC101927472 |
Gene Aliases: |
- |
Location: |
Chr.10:104323364-104326745 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927472 | NR_120622.1 | |||
| BM794484.1 | ||||
| BX094877.1 |

Genomic Map