Assay Details
Target Gene Details
Entrez Gene ID: | 253430 |
Gene Name: | inositol polyphosphate multikinase |
Gene Aliases: |
- |
Location: |
Chr.10:58191517-58269837 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPMK | NM_152230.4 | NP_689416.1 | ||
| XM_011539565.2 | XP_011537867.1 | |||
| XM_017016013.1 | XP_016871502.1 | |||
| AF432853.1 | AAM97838.1 | |||
| BC065709.1 | AAH65709.1 |
More Information
Set Membership: |
|

Genomic Map