Assay Details
Target Gene Details
Entrez Gene ID: | 9124 |
Gene Name: | PDZ and LIM domain 1 |
Gene Aliases: |
CLIM1, CLP-36, CLP36, HEL-S-112, hCLIM1 |
Location: |
Chr.10:95237568-95291148 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDLIM1 | NM_020992.3 | NP_066272.1 | ||
| XM_011540330.1 | XP_011538632.1 | |||
| AJ310549.1 | CAC32846.1 | |||
| AK314792.1 | ||||
| BC000915.2 | AAH00915.1 | |||
| BC018755.1 | AAH18755.1 | |||
| FJ224331.1 | ||||
| U90878.1 | AAC05580.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv22688 | Chr.10:95096249 - 95314999 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map