Assay Details
Target Gene Details
Entrez Gene ID: | 23234 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C9 |
Gene Aliases: |
HDJC9, JDD1, SB73 |
Location: |
Chr.10:73241954-73247331 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC9 | NM_015190.4 | NP_056005.1 | ||
| AF327347.1 | AAL56008.1 | |||
| AK074420.1 | BAB85076.1 | |||
| AK094162.1 | ||||
| BC136507.1 | ||||
| BX646217.1 |
More Information
Set Membership: |
|

Genomic Map