Assay Details
Target Gene Details
Entrez Gene ID: | 5407 |
Gene Name: | pancreatic lipase related protein 1 |
Gene Aliases: |
PLRP1 |
Location: |
Chr.10:116590415-116609175 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNLIPRP1 | NM_001303135.1 | NP_001290064.1 | ||
| NM_006229.3 | NP_006220.1 | |||
| XM_011539868.1 | XP_011538170.1 | |||
| XM_011539869.1 | XP_011538171.1 | |||
| AK130236.1 | ||||
| BC025784.2 | AAH25784.1 | |||
| CD622211.1 | ||||
| CR749299.1 | CAH18154.1 | |||
| CR749524.1 | CAH18337.1 | |||
| CR749596.1 | CAH18394.1 | |||
| M93283.1 | AAA59532.1 |
More Information
Set Membership: |
|

Genomic Map