Assay Details
Target Gene Details
Entrez Gene ID: | 5588 |
Gene Name: | protein kinase C theta |
Gene Aliases: |
PRKCT, nPKC-theta |
Location: |
Chr.10:6393038-6580646 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCQ | NM_001323267.1 | NP_001310196.1 | ||
| XM_005252497.4 | XP_005252554.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053177 | Chr.10:6411345 - 6431152 on Build GRCh38 | Loss |
|
| nsv1161925 | Chr.10:6417656 - 6429854 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map