Assay Details
Target Gene Details
Entrez Gene ID: | 840 |
Gene Name: | caspase 7 |
Gene Aliases: |
CASP-7, CMH-1, ICE-LAP3, LICE2, MCH3 |
Location: |
Chr.10:113679162-113730909 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASP7 | NM_001227.4 | NP_001218.1 | ||
| NM_001267056.1 | NP_001253985.1 | |||
| NM_001267057.1 | NP_001253986.1 | |||
| NM_001320911.1 | NP_001307840.1 | |||
| NM_033338.5 | NP_203124.1 | |||
| NM_033339.4 | NP_203125.1 | |||
| NM_033340.3 | NP_203126.1 | |||
| XM_006718017.3 | XP_006718080.1 | |||
| XM_017016763.1 | XP_016872252.1 | |||
| XM_017016764.1 | XP_016872253.1 | |||
| AB451281.1 | ||||
| AB451413.1 | ||||
| AK223489.1 | ||||
| AK300578.1 | ||||
| AK301437.1 | ||||
| AU130224.1 | ||||
| BC015799.1 | AAH15799.1 | |||
| BX487250.1 | ||||
| BX647973.1 | ||||
| DC364916.1 | ||||
| DC375817.1 | ||||
| DC384804.1 | ||||
| DC405356.1 | ||||
| DC407017.1 | ||||
| U37448.1 | AAC50303.1 | |||
| U37449.1 | AAC50304.1 | |||
| U40281.1 | AAC50352.1 | |||
| U67206.1 | AAF21460.1 | |||
| U67320.1 | AAC51153.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624603 | Chr.10:113581857 - 113745416 on Build GRCh38 | Gain |
|
| nsv1048819 | Chr.10:113499347 - 113757619 on Build GRCh38 | Gain |
|
| esv3624607 | Chr.10:113679337 - 113732388 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map