Assay Details
Target Gene Details
Entrez Gene ID: | 6840 |
Gene Name: | supervillin |
Gene Aliases: |
- |
Location: |
Chr.10:29457338-29737001 on Build GRCh38 |
Assay Gene Location: | Within Intron 31 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038080 | Chr.10:29361590 - 29565510 on Build GRCh38 | Gain |
|
| nsv519798 | Chr.10:29332490 - 29500165 on Build GRCh38 | Loss |
|
| nsv6210 | Chr.10:29481156 - 29503058 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map