Assay Details
Target Gene Details
Entrez Gene ID: | 5241 |
Gene Name: | progesterone receptor |
Gene Aliases: |
NR3C3, PR |
Location: |
Chr.11:101029624-101130681 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGR | NM_000926.4 | NP_000917.3 | ||
| NM_001202474.3 | NP_001189403.1 | |||
| NM_001271161.2 | NP_001258090.1 | |||
| NM_001271162.1 | NP_001258091.1 | |||
| NR_073141.2 | ||||
| NR_073142.2 | ||||
| NR_073143.2 | ||||
| XM_006718858.3 | XP_006718921.1 | |||
| AB084248.1 | BAB91074.1 | |||
| AB085683.1 | BAC06585.1 | |||
| AB085843.1 | BAC11011.1 | |||
| AB085844.1 | BAC11012.1 | |||
| AB085845.1 | BAC11013.1 | |||
| AF016381.1 | AAD01587.1 | |||
| AK304853.1 | ||||
| AY212933.1 | 1 | 1029 | AAO61671.1 | |
| AY382151.1 | AAQ96833.1 | |||
| M15716.1 | AAA60081.1 | |||
| X51730.1 | CAA36018.1 |
More Information
Set Membership: |
|

Genomic Map