Assay Details
Target Gene Details
Entrez Gene ID: | 159963 |
Gene Name: | solute carrier family 5 member 12 |
Gene Aliases: |
SMCT2 |
Location: |
Chr.11:26667019-26723356 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A12 | NM_178498.3 | 15 | 3483 | NP_848593.2 |
| XM_006718155.3 | 15 | 3386 | XP_006718218.1 | |
| XM_011519920.2 | 16 | 2963 | XP_011518222.1 | |
| XM_017017244.1 | 16 | 3409 | XP_016872733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819331 | Chr.11:26669621 - 26669831 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map