Assay Details
Target Gene Details
Entrez Gene ID: | 341152 |
Gene Name: | olfactory receptor family 2 subfamily AT member 4 |
Gene Aliases: |
OR11-265 |
Location: |
Chr.11:75088692-75100854 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR2AT4 | XM_017017655.1 | 2 | 1132 | XP_016873144.1 |
| XM_017017656.1 | 2 | 1128 | XP_016873145.1 | |
| XM_017017657.1 | 2 | 670 | XP_016873146.1 | |
| XM_017017658.1 | 2 | 1155 | XP_016873147.1 |
More Information
Set Membership: |
|

Genomic Map