Assay Details
Target Gene Details
Entrez Gene ID: | 23682 |
Gene Name: | RAB38, member RAS oncogene family |
Gene Aliases: |
NY-MEL-1, rrGTPbp |
Location: |
Chr.11:87809490-88175505 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB38 | XM_017017456.1 | XP_016872945.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1249n100 | Chr.11:87802086 - 88022773 on Build GRCh38 | Gain |
|
| nsv1042017 | Chr.11:87152177 - 88155066 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map