Assay Details
Target Gene Details
Entrez Gene ID: | 56946 |
Gene Name: | EMSY, BRCA2 interacting transcriptional repressor |
Gene Aliases: |
C11orf30, GL002 |
Location: |
Chr.11:76444928-76552901 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMSY | NM_001300942.1 | NP_001287871.1 | ||
| NM_001300943.1 | NP_001287872.1 | |||
| NM_001300944.1 | NP_001287873.1 | |||
| NM_020193.4 | NP_064578.2 | |||
| XM_005274112.4 | XP_005274169.1 | |||
| XM_006718637.3 | XP_006718700.1 | |||
| XM_006718638.3 | XP_006718701.1 | |||
| XM_006718639.3 | XP_006718702.1 | |||
| XM_011545173.2 | XP_011543475.1 | |||
| XM_011545174.2 | XP_011543476.1 | |||
| XM_011545175.2 | XP_011543477.1 | |||
| XM_011545177.2 | XP_011543479.1 | |||
| XM_011545178.2 | XP_011543480.1 | |||
| XM_011545179.2 | XP_011543481.1 | |||
| XM_017018021.1 | XP_016873510.1 | |||
| AK125114.1 | ||||
| AK304043.1 | ||||
| AK309621.1 | ||||
| BC033404.1 | AAH33404.1 | |||
| BG722092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050713 | Chr.11:76436041 - 76556073 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map