Assay Details
Target Gene Details
Entrez Gene ID: | 374407 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B13 |
Gene Aliases: |
RSPH16A, TSARG5, TSARG6 |
Location: |
Chr.11:73950319-73970287 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB13 | NM_153614.3 | NP_705842.2 | ||
| XM_005273984.3 | XP_005274041.1 | |||
| XM_011545004.2 | 2 | 661 | XP_011543306.1 | |
| XM_011545005.2 | 2 | 663 | XP_011543307.1 | |
| XM_011545009.2 | 2 | 494 | XP_011543311.1 | |
| XM_011545013.2 | 2 | 667 | XP_011543315.1 | |
| XM_011545014.2 | 2 | 667 | XP_011543316.1 | |
| XM_017017679.1 | 2 | 667 | XP_016873168.1 | |
| AK128589.1 | ||||
| AY138810.2 | AAN15929.1 | |||
| AY325766.1 |
More Information
Set Membership: |
|

Genomic Map