Assay Details
Target Gene Details
Entrez Gene ID: | 116534 |
Gene Name: | MAS related GPR family member E |
Gene Aliases: |
GPR167, MGRF, MRGE |
Location: |
Chr.11:3227811-3232386 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRGPRE | NM_001039165.2 | 1 | 119 | NP_001034254.2 |
| AK293940.1 | 1 | 119 | ||
| DC309261.1 | 1 | 119 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759796 | Chr.11:3215282 - 3780199 on Build GRCh38 | Gain+Loss |
|
| nsv951295 | Chr.11:3213271 - 3236070 on Build GRCh38 | Deletion |
|
| dgv176e214 | Chr.11:3217520 - 3339788 on Build GRCh38 | Gain |
|
| nsv7649 | Chr.11:3211044 - 3362675 on Build GRCh38 | Deletion |
|
| esv3891908 | Chr.11:3220993 - 3381247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map